Learn More
Enzo Life Sciences Cytochalasin D, (1mg), CAS Number: 22144-77-0
Supplier: Enzo Life Sciences BMLT1090001
Ultra-pure; inhibits actin polymerization; K.+ channels. Modulates CD4 cross-linking in T-lymphocytes and increases intracellular Ca2+ levels. Exhibits antitumor activity. Induces apoptosis. Formula: C30H37NO6. MW: 507.6. CAS Number: 22144-77-0. Purity: ≥97% (HPLC, single spot by TLC). Appearance: White to off-white solid. Solubility: Soluble in methylene chloride (10mg/ml) or DMSO (up to 25mg/ml). Long Term Storage: -20°C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.