Learn More
Bioworld Cytidine, 25 g
Supplier:  Bioworld 403900041
Encompass_Preferred
It is a nucleoside molecule that is formed when cytosine is attached to a ribose ring (also known as a ribofuranose) . Cytidine is a component of RNA.If cytosine is attached to a deoxyribose ring, it is known as a deoxycytidine.Dietary sources of cytidine include foods with high RNA (ribonucleic acid) contents such as organ meats, Brewer's yeast, as well as pyrimide-rich foods such as beer. During digestion, RNA-rich foods are broken-down into ribosyl pyrimidines (cytidine and uridine), which are absorbed intactIn humans, dietary cytidine is converted into uridine,which is probably the compound behind cytidine's metabolic effects.Specifications:Em (280nm, 0.1N HCl)- >12600Ultra Pure Grade Appearance: White crystalline powderBoiling Point : 545.7 °CChEBI : Not available.Crystal_Structure : Not available.Density : 1.89 g/cm3InChI : Not available.InChIKey : Not available.Melting Point: 210-220°CRTECS Number: UW7370000
Catalog No. 50-255-840