Learn More
Kamiya Biomedical Cystatin C Reagent, 20mL+20mL/PK RX
Supplier: Kamiya Biomedical KAI074
K-ASSAY Cystatin C Assay Reagent (L). R1: 2 x 10mL (liquid), R2: 2 x 10mL (liquid). Assay Range: 0.40 - 8.00 mg/L (0.34 - 6.80 mg/L ERM-DA471/IFCC Standardized). The K-ASSAY Cystatin C assay is intended for the quantitative determination of human cystatin C in serum, EDTA plasma, or lithium heparin plasma by immunoturbidimetric assay. Cystatin C measurements are used as an aid in the diagnosis and treatment of renal diseases. FOR IN VITRO DIAGNOSTIC USE. Required Calibrator: K-ASSAY Cystatin C Calibrator (Cat. No. KAI-099C). Recommended Controls: K-ASSAY Cystatin C Control (Cat. No. K100C-2M or K100C-4M). This assay is intended for use on most popular chemistry analyzers that allow open channel reagents. Please contact KAMIYA BIOMEDICAL COMPANY to check parameter availability for your analyzer.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.