Learn More
Kamiya Biomedical Cystatin C Calibrator, 6x1mL/PK RX
Supplier: Kamiya Biomedical KAI099C
K-ASSAY Cystatin C Calibrator. 6 x 1 mL (liquid). For 6 point calibration. The K-ASSAY Cystatin C Caibrator is intended to be used for the calibration of the K-ASSAY Cystatin C immunoturbidimetric assay (Cat. No. KAI-073 or KAI-074). FOR IN VITRO DIAGNOSTIC USE.