Learn More
CPC Scientific H-Cys(Acm)-Met-His-Ile-Glu-Ser-Leu-Asp-Ser-Tyr-Thr-Cys(Acm)-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific GRFF001B
Encompass
ONE-LETTER SEQUENCE: Cys(Acm)-MHIESLDSYT-Cys(Acm)MOLECULAR FORMULA: C63H98N16O23S3MOLECULAR WEIGHT:1543.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [89991-90-2], RESEARCH AREA: CancerREFERENCES: A. Komoriya et al., PNAS, 81, 1351 (1984)
Catalog No. 50-210-9393
Explore available promotions