Learn More
PBL Assay Science VeriKine Cynomolgus/Rhesus Interferon Alpha (IFN-Alpha, IFNa) ELISA [1x96 well plate]
Supplier: PBL Assay Science 461001
VeriKine Cynomolgus/Rhesus Interferon Alpha Serum ELISA is manufactured by PBL Assay Science (Catalog #46100-1) for research use only (RUO) and packaged as a 1x96-well plate. This assay is serum, plasma, and tissue culture media (TCM) compatible and has an assay range of 25-1,600 pg/mL. This ELISA kit has intra- and inter-assay CVs of less than 10%.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.