Learn More
Electron Microscopy Sciences JEOL SEM Cylinder Mount 1-1/4" surface, 3/16" height, Aluminum
Supplier: Electron Microscopy Sciences 7583040
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 1-1/4"
Height: 3/16"