Learn More
Hach Company Cylinder, Graduated, Mixing, Glass, 50 mL +-0.4 mL, 1.0mL divisions, glass stopper #16
Supplier: Hach Company 189641
Scale is durable white ceramic enamel. Designed to meet ASTM Specification E1272, Style 2 (Class B). Includes glass stopper.