Learn More
Hach Company Cylinder, Graduated, Mixing, Glass, 10 mL +-0.1 mL, 0.2 mL divisions, glass stopper #13
Supplier: Hach Company 2088638
This glass graduated cylinder is calibrated “to contain” with a molded in base. Each includes glass penny head stopper. Graduations are in durable white enamel bake.