Learn More
Perkin Elmer US LLC Standard Cyclonic Spray Chamber for Optima 2x00/4x00/5x00/7x00/8x00
Supplier: Perkin Elmer US LLC N0776052
Standard and 0-Ring Free glass cyclonic spray chambers can be used with either GemCone or concentric glass nebulizers. HF resistant cyclonic spray chambers should be used with GemCone, SeaSpray or MicroMist nebulizers. This spray chamber is included in the Cyclonic/Concentric Sample Introduction Kit for Optima 8x00 (N0780617) and the Cyclonic/Concentric Sample Introduction Kit for Optima 2x00/4x00/5x00/7x00 DV (N0770730). The Standard Cyclonic Spray Chamber Nebulizer Adapter Plug (N0776006) available separately.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.