Learn More
Bachem Cyclo(-Ala-Gln)
Supplier: Bachem 4035535.1000
Cyclo(-Ala-Gln), 268221-76-7, C8H13N3O3, Mr 199.21, 1g. Cyclo(Ala-Gln) is a potential impurity of L-alanyl-L-glutamine used for glutamine supplementation.