Learn More
Evident Crime Scene Products CYANOACRYLATE GLUE - 16 OZ
SDP
Supplier:  Evident Crime Scene Products 3027EV
Encompass_Preferred
his large 16 oz. Cyanoacrylate is well suited for all high volume processing requirements. The Cyanoacrylate may be applied directly from the bottle, or refilled into smaller bottles. Simply heat the Cyanoacrylate with an aluminum dish on either the MicroBurst Variable Glue Heating Plate or the standard Glue Heating Plate to produce fumes. Always wear a respirator, goggles, and gloves.
Catalog No. NC2583408