Learn More
Chemglass Life Sciences Electroporation Cuvettes, 1mm
SDP
Supplier:  Chemglass Life Sciences CLS137001
Encompass_Preferred
Electroporation Cuvettes, 1mm. Bridge Electroporation Cuvettes efficiently transfer DNA and siRNA into a wide variety of cell types. While the color and shape of the caps may vary from brand to brand, all electroporation cuvettes must meet strict quality assurance specifications to ensure consistent pulse delivery and reproducible results. Bridge cuvettes meet and exceed every measurable qualification and parameter. It is well documented that nucleic acid delivery via electroporation can achieve remarkable rates of transfer efficiency and maintain good cell viability, even for a variety of finicky human cell lines. In order to achieve the proper electrical field strength to the cells, Bridge Electroporation Cuvettes are carefully constructed with burr-free, specially processed electrodes and virgin polycarbonate. This results in cuvettes which provide superior results in the most discriminating applications.
Catalog No. 50-153-2109