Learn More
Electron Microscopy Sciences Curved Forceps, 153 mm OAL
Supplier: Electron Microscopy Sciences 72910
Curved forceps are made of stainless steel and have medium fine tips with curved smooth points.
OAL: 153 mm