Learn More
Pharmacal Research Laboratories CURTIS CYCLONE ULTRA FOGGER
SDP
Supplier:  Pharmacal Research Laboratories 990501
Encompass
model 2736220/240v ac 4.2amps 50/60hz ,flow rate:0-2.4oz(70ml./min.)via metering valve, droplet size 5-20 microns, tank capacity:1 gallon (3.8 liters)
Catalog No. NC2084708