Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD CultureSwab™ MaxV and MaxV+ Collection and Transport Systems
Improved recovery of fastidious organisms for proper patient diagnosis
Supplier: BD 220232
Description
- Improved recovery of fastidious organisms such as N. gonorrhoeae, H. influenzae, N. meningitidis, and S. pneumoniae
- Sterile, ready-to-use systems intended for collection, transport, and preservation of clinical specimens for bacteriological examination
- Medium is designed to maintain viability of organisms during transit to the laboratory
- Pouches are made of a plastic film which retards penetration of atmospheric air into the product
- Nitrogen-flushed packaging
- Patented tube design
- Tamper-evident seal
- Double-action security cap
- Shaft: 5 1/4 in.
Specifications
Collection and Transport System | |
Sterile | |
Collection, transport and preservation of clinical specimens for Bacteriological examination | |
50/Pk. |
MaxV Liquid Amies | |
Dual Applicator | |
Nitrogen flushed | |
5°C to 25°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction