Learn More
CELLTREAT Scientific Products 14mL Culture Tube and Dual Cap, PS, Sterile
Supplier: CELLTREAT Scientific Products 230440
Dual caps allow for either vented or sealed positions. Polystyrene tubes are optically clear to visualize cultures. Suitable for FACS and flow cytometry applications. Used for bacterial & suspension culture.
This item is not returnable.
View return policy