Learn More
CELLTREAT Scientific Products 14mL Culture Tube and Dual Cap, PP, Sterile
Supplier: CELLTREAT Scientific Products 230439
Dual caps allow for either vented or sealed positions.14mL tubes feature molded graduations. Polypropylene tubes offer better chemical resistance. Suitable for FACS and flow cytometry applications. Used for bacterial & suspension culture.
This item is not returnable.
View return policy