Learn More
R&D Systems™ Cultrex Organoid Harvesting Solution
Depolymerizes basement membrane matrix for harvesting organoids from culture.
Supplier: R&D Systems™ 370010001
Description
Embryonic stem (ES) cells are pluripotent stem cells derived from the inner cell mass of pre-implantation embryos. Induced pluripotent stem (iPS) cells can be generated by somatic cell reprogramming following the exogenous expression of specific transcription factors (Oct-3/4, KLF4, SOX2, and c-Myc). These cell types are capable of unlimited, undifferentiated proliferation in vitro and still maintain the capacity to differentiate into a wide variety of somatic cells. In this capacity, pluripotent stem cells have widespread clinical potential for the treatments of heart disease, diabetes, spinal cord injury, and a variety of neurodegenerative disorders.
Applications: Depolymerizes basement membrane matrix for harvesting organoids from culture
Storage: Store unopened product at 2° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Specifications
Store the unopened product at 2 - 8° C. Do not use past expiration date. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only