Learn More
Thermo Scientific™ Culti-Loops™ Escherichia coli serotype O157:H7 ATCC™ 43888™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Supplier: Thermo Scientific™ R4601965
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
43888TM* | |
Culti-Loops | |
Gram-negative Rods | |
CDC B6914-MS1 |
Escherichia coli serotype O157:H7 ATCC™ 43888™; Does not produce or possess genes for Shiga-like toxin I or II. | |
Culti-Loop | |
5 Loops/Pack |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.