Learn More
Sartorius Cubis™ II Semi-Micro Balance, MCE User Interface
Cubis™ II Semi-Micro Balance features fully customizable hardware, software, and connectivity
Supplier:  Sartorius MCE225SUS00
Request Bulk Order
 View more versions of this product
Catalog No. 14-561-267
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Explore available promotions
Description
Description
Cubis™ II Semi-Micro Balance features fully customizable hardware, software, and connectivity
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor (draft shield M), or gesture sensor (optional) learning capability
Warranty and Services
10-year warranty
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
Color TFT Touchscreen
Balance, Accessories, Power supply, Manual, Factory test report; QP99 software
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
± 0.01 mg
Semi-micro Balance
404 mm
373/61 mm weighing/electrical module
120/240 V
50/60 Hz
220 g
0.01 mg
± 0.065 mg
Draft shield U: Manual glass
2 sec.
10 years
240 mm
10.2, 11.7 kg
100/240 V ±10%
Product Suggestions
Product Suggestions