Learn More
Medchemexpress LLC HY-101929 5mg Medchemexpress, CU-CPT17e CAS:2109805-75-4 Purity:>98%
Supplier: Medchemexpress LLC HY1019295MG
Medchemexpress, HY-101929 5mg CU-CPT17e CAS:2109805-75-4 CU-CPT17e is a potent multi-Toll-like receptor (TLR) agonist that activates TLR3, TLR8, and TLR9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.