Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ad Infinitum™ CryoProtectPURE™ Cryopreservation Media PROMO
Ad Infinitum™ CryoProtectPURE™ Cryopreservation Media is a DMSO-free and Serum-free cryopreservation media for cell and tissue preservation.
Supplier: Ad Infinitum™ CPP100100PM
Description
- Increases the cell viability during freeze and post-thaw, producing healthier cells with no antigenicity concern or toxic vitrification.
- Serum-free
- Pre-mixed
- Ready-to-use
- Filtered to 0.2 μm
- Suitable for a wide variety of cells
- Shipped at ambient temperature
- For manufacturing and/or research use only, not for diagnostic or therapeutic use
- Manufactured at a cGMP-compliant facility registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards
Specifications
ISO 13485 standards | |
Pre-mixed and ready-to-use | |
Manufacturing and/or Research Use Only, Not for Diagnostic or Therapeutic Use |
2°C to 8°C | |
Cryopreservation Media | |
100 mL |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction