Learn More
Azenta US Inc CryoPod™ Carrier
Supplier: Azenta US Inc 243354001
The LN2 vapor-based CryoPod™ Carrier provides a safe, portable, and trackable solution for hand carrying temperature-sensitive biological materials.Holds samples at ≤-150°C for over 3 hours. Displays and logs temperature, date, time. Audible and visual temperature alarms. Compact, lightweight, portable.
This item is not returnable.
View return policy