Learn More
Molecular Dimensions CRYOMOUNT SET 0.2MM
SDP
Supplier:  Molecular Dimensions MD7136CM
Encompass_Preferred
Protein Crystallography. A set of mounted 0.20 mm LithoLoops mounted in CryoCaps and Magnetic CryoVials. Each CryoCap is DataMatrix etched with the size of the LithoLoop. The sets are provided in a handy storage box.
Catalog No. NC0071662