Learn More
Cole-Parmer Corning 430488 Cryogenic Vials, Internal Thread/Washer/Self-Stand/2.0 mL; 50/Pk
Supplier: Cole-Parmer 4435112
Vial caps will hold Corning color-coded inserts. Lock into Corning cryogenic racks for single-handed operation. Cap inserts and self-locking racks are sold separately below right. Note: Do not autoclave or immerse vials in liquid nitrogen. For storage under extreme conditions such as liquid nitrogen, use CryoFlex™ tubing 03755-64.