Learn More
Taylor Wharton CRYO REFRIGERATOR HC20
Supplier: Taylor Wharton 20HCB11M
Refrigerator, Static Holding Time: 87days, Working Time: 54days, Evaporation Rate: 0.23L/days, Liquid Nitrogen Capacity: 20L, Neck Diameter: 3.6 in., Overall Height: 24.25 in., Overall Diameter: 15.6 in., Number of Canisters: 6