Learn More
Kamiya Biomedical CRP Calibrator, 6x1mL/PK
Supplier: Kamiya Biomedical KAI012C
K-ASSAY CRP Calibrator. 6 x 1 mL (liquid). For 6 point calibration. The K-ASSAY CRP Calibrator is intended to be used for the calibration of the K-ASSAY CRP immunoturbidimetric assay (Cat. No. KAI-026). FOR IN VITRO DIAGNOSTIC USE.