Learn More
United Scientific Supplies Cross-Shaped Magnetic Stir Bars
High quality magnets coated with PTFE
$10.50
Specifications
Autoclavable | Autoclavable |
---|---|
Color | White |
Material | PTFE Coated Magnet |
Shape | Cross |
Description
- High-quality magnets coated with durable PTFE
- Cross shape increases turbulence and improves stirring action
- Available in 2 sizes
Specifications
Autoclavable | |
White | |
PTFE Coated Magnet | |
Cross |