Learn More
Crime Scene Lab Investigations
Scenario transforms students into forensic scientists.
Supplier: Neo/Sci Corporation 203863
Includes: All supplies needed to set up a variety of scenarios
Description
Gather evidence at the crime scene; analyze it at the “crime lab.”
- Provides suggestions on integrating the skills learned into your curriculum
- Examine physical evidence such as hair, fiber, shoe prints, blood, chromosomes and DNA, and tool marks
- Perform tests to analyze evidence collected
- Identify the crime victim through dental records and fingerprints
- Perform tests to determine cause of death
- Analyze evidence to determine a suspect
- Class size: 40 students
Specifications
Crime Scene Investigations Lab Investigation Kit | |
For scenario transforms students into forensic scientists | |
All supplies needed to set up a variety of scenarios | |
40 Students |