Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Certified Safety CPRotector™ 2000
Prevents unsafe mouth-to-mouth contact
Specifications
Material | HDPE |
---|---|
Disposable | Yes |
For Use With (Application) | Prevents unsafe mouth-to-mouth contact |
Catalog Number | Mfr. No. | Color | Product Type | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Product Type | Includes | Price | Quantity & Availability | |||
19-822-043
|
Certified Safety
216068 |
Clear | CPR Barrier | Comprehensive instructions | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||
Description
- Disposable, single-use mouth-to-mouth barriers provide protection to rescuers from victim's oral secretions
- One-way valve allows maximum amount of air to be sent to victim while preventing return of air or body fluids to rescuer
- Cross-section gate prevents tongue blockage
- Gate can also be used as a bite stick
- Patented design is made of clear, high-density polypropylene
- Remains soft and flexible to -10°F (-23°C)
- Instructions for use printed on unit
Order Info
Available packaged in a light smoke poly bag with a pair of latex gloves or without gloves in a loose-fit, easy-to-open clear overwrap.
Compliance
FDA 510(K) registered
Specifications
HDPE | |
Yes | |
Prevents unsafe mouth-to-mouth contact |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title