Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPPG
Potent group III mGlu antagonist
$149.64 - $560.28
Specifications
Chemical Name or Material | (RS)-α-Cyclopropyl-4-phosphonophenylglycine |
---|---|
CAS | 183364-82-1 |
Assay Percent Range | >98% |
Target | Group III mGluR Antagonists |
Molecular Formula | C11H14NO5P |
Description
Potent group II/III mGlu receptor antagonist, with approximately 20-fold selectivity for group III over group II (IC50 values of 2.2 and 46.2 nM respectively). A much less potent antagonist at group I receptors in neonatal rat cortical slices (KB = 0.65 ± 0.07 nM).Specifications
(RS)-α-Cyclopropyl-4-phosphonophenylglycine | |
>98% | |
C11H14NO5P | |
98% |
183364-82-1 | |
Group III mGluR Antagonists | |
Observed MW: 271.21 |
Safety and Handling
Recommended Storage : Store at Room Temperature
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title