Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix Coxsackievirus Type A9 Culture Fluid
Infectious virus preparation designed to meet research and product development needs
Supplier: ZeptoMetrix 0810017CF
Alert:
Contains live and infectious organisms.
Description
Coxsackievirus is a non-enveloped enterovirus that contains a linear, positive-sense single-stranded RNA.
- Consist of virus, cells and media taken directly from tissue culture flask
- Propagated in Vero cell line
- Biosafety Level 2 organism
- Available in 1.0mL aliquots
- Each lot is is assayed for its Tissue Culture Infective Dose (TCID50)
- Titer: >105 U/mL
IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.
Nucleic Acid/Molecular Testing, Limit of Detection (LOD) Studies, Cross-eactivity Studies, Other Viral-based Assays
Specifications
2 | |
>105U/mL | |
Solution | |
Coxsackievirus Infectious Culture Fluid | |
1 mL |
ISO, 13485 | |
-65°C | |
Vial | |
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays |
Safety and Handling
ShelfLife : 1 Year
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction