Learn More
Covidien Monoject™ SoftPack Tuberculin Syringes
Sterile, single-use individually packaged syringes
Specifications
Latex Free Yes
Product Type Softpack Tuberculin Syringe
Disposable Yes
Volume (Metric) 1 mL
Graduated Yes
View More Specs
Products 7
Catalog Number Mfr. No. Connection Type Needle Gauge Price Quantity & Availability  
Catalog Number Mfr. No. Connection Type Needle Gauge Price Quantity & Availability  
22-257-135
View Documents
Covidien
1180100555
Luer
Pack of 100 for $36.70
Case of 5 PK for $156.20
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-257-136
View Documents
Covidien
1180125058
Removable 25 G
Pack of 100 for $33.40
Case of 5 PK for $141.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-257-137
View Documents
Covidien
1180125158
Fixed 25 G
Pack of 100 for $38.00
Case of 5 PK for $160.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-257-138
View Documents
Covidien
1180126038
Detachable 26 G
Case of 500 for $165.20
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-257-139
View Documents
Covidien
1180127012
Detachable 27 G
Pack of 100 for $33.60
Case of 5 PK for $142.99
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-257-140
View Documents
Covidien
1180128012
Fixed 28 G
Pack of 100 for $38.00
Case of 5 PK for $160.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-257-150
View Documents
Covidien
1180100777
Luer Tip
Case of 240 for $466.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Polypropylene barrel and plunger rod
  • Latex-free plunger tip
  • Graduated barrel markings
  • Variety of needle gauge sizes
Specifications
Specifications
Yes
Yes
Yes
Softpack Tuberculin Syringe
1 mL
Sterile