Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Covidien Dermacea™ 100% Cotton Gauze Sponges
An alternative for sterile and nonsterile 100% cotton gauze dressings
Supplier: Covidien 442308
Specifications
Cotton | |
Non-sterile | |
2 x 2 in. | |
8-Ply |
Gauze Sponge | |
5000 Cs. | |
Wound dressing, general cleaning, minor prepping, wound packing and debriding |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction