Learn More
Revvity Health Sciences Inc COUNT-OFF Liquid Concentrate; 4x2.5L
Supplier:  Revvity Health Sciences Inc 6NE9427
Encompass_Preferred
Ideal, all-purpose decontaminant: safe, mild, efficient and economical. Quickly and effectively cleans up even the most persistent leftovers: proven most effective cleanser for removing radioactive residues (3H, 14C, 32P and 125I), stopcock and vacuum greases, lanolin and petroleum jelly, dried blood and serum, fatty and amino acids, protein complexes, and polymer films and other stubborn residues Stable under extreme temperature fluctuations (-50 'C to 150 'C) Safer than strongly acidic cleaners, such as chromic acid: solutions will not produce toxic gases from substrates containing 14C, 131I, 35S, or 36Cl Easy to use: adjust concentration, soaking time and temperature as needed to speed up decontamination or for particularly stubborn substances such as dried blood
Catalog No. 50-905-1212