Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coumarin 504T 98.0+%, TCI America™
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Click to view available options
Quantity:
200 mg
1 g
Specifications
Melting Point | 129°C |
Quantity | 1 g |
Formula Weight | 369.46 |
Percent Purity | ≥98.0% (HPLC,N) |
Physical Form | Crystalline Powder |
Chemical Name or Material | Coumarin 504T |
Safety and Handling
TSCA | No |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction