Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Cotton Twine for Science Kits

Consumable used as a science kit component that is compatible with multiple projects and kits.
Description
- Cotton
- Medium weight
- 420 ft.
Specifications
Medium Weight Twine | |
For multiple projects or in a science kit | |
128 m |
Cotton | |
420 ft. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title