Learn More
Gibco™ Cotton Peptone 200, ultra-filtered (UF)
Gibco™ Cotton Peptone 200, Ultra-Filtered (UF) is processed to enhance carbohydrate concentrations, which is important for some mammalian protein glycosylation processes.
Supplier: Gibco™ 670104
Description
Cotton 200, UF, is also part of the Peptone Preview Pak which contains:
- Cotton 100, UF—rich in nucleoside content and digested to enhance peptide content. It is processed to provide higher concentrations of vitamins, which can help maintain performance in mammalian and microbial processes.
- Wheat 100, UF—ultra-filtered supplement and good source of glutamine, which makes it ideal for mammalian cell culture applications. It is digested for enhanced peptide content resulting in a low concentration of free amino acids and carbohydrates.
- Soy 100—highly digested soy protein peptone that is high in nucleosides and free amino acid content. It has high concentrations of vitamins and carbohydrates and is recommend for a wide variety of processes.
- Pea 100, UF—ultra-filtered peptone digested to enhance nucleoside and free amino acid content. The high total amino acid content and low carbohydrate content are similar to many high-performance animal origin peptones.
- Yeast 100—designed to enhance the content of vitamins and other key nutritional components found in yeast. It is intended for nutritional supplementation for mammalian, microbial, and insect cell culture.
Peptone Preview Pak supplements are non-GMP prototype products that are not for manufacture use. Larger sizes are available through the Gibco Custom Products Services.
Specifications
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. | |
Powder | |
Mammalian Cell Culture | |
Animal origin-free |
Mammalian, Insect, Microbial | |
Dehydrated Culture Media | |
500 g |
Safety and Handling
ShelfLife : 2 Years
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.