Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ Filtered IsoTip™ Universal Fit Racked Pipet Tips
Ideal for applications where avoiding cross-contamination is critical, such as DNA amplification and radioisotope handling
$230.00 - $396.00
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Color | Natural |
Format | Rack |
Tip Style | Round |
Includes: Pipet tips in rack
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
07-200-502
|
Corning™
4808 |
0.5 to 10 μL |
Case of 960 for $396.00
|
|
|||||
07-200-308
|
Corning™
4801 |
0.1 to 2 μL |
Case of 960 for $252.00
|
|
|||||
07-200-265
|
Corning™
4821 |
1 to 30 μL |
Case of 960 for $230.00
|
|
|||||
07-200-501
|
Corning™
4807 |
0.2 to 10 μL |
Case of 960 for $252.00
|
|
|||||
07-200-503
|
Corning™
4809 |
100 to 1000 μL |
Case of 1000 for $291.00
|
|
|||||
07-200-267
|
Corning™
4823 |
1 to 200 μL |
Case of 960 for $230.00
|
|
|||||
Description
- IsoTip filtered pipet tips feature an inert, hydrophobic barrier that prevents aerosolized contaminants from coming in contact with pipettor shafts. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website.)
- Ideal for applications where avoiding cross contamination is critical, such as DNA amplification and radioisotope handling.
- Packaged sterile.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifications
Filtered | |
Natural | |
Round | |
Isotip |
Sterile | |
Rack | |
Rack | |
Polypropylene |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title