Learn More
R&D Systems™ Corning™ Costar™ Spin-X™ Centrifuge Tube Filters
Cellulose acetate membrane
Specifications
Description | Cellulose acetate (CA) membranes with 0.22 micron pore size |
---|---|
Content And Storage | Store the unopened product at room temperature. Do not use past expiration date. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
SPNX8160016
|
R&D Systems™
SPINX8160016 |
16 | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Filters by centrifugation for removing bacteria, particles, or cells from liquids, HPLC sample preparation and DNA removal from agarose or acrylamide gels
- Cellulose acetate (CA) membranes provide fast flow rates and low protein binding
- Pore size 0.22 micron
- Sterile
Specifications
Cellulose acetate (CA) membranes with 0.22 micron pore size | |
Store the unopened product at room temperature. Do not use past expiration date. |