Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Cork Stoppers
SDPSCI_ED

Grade XXX Tapered Cork Stoppers are an excellent choice for general laboratory use

Supplier:  United Scientific Supplies CST1

 View more versions of this product

Catalog No. S39360


Add to Cart

Description

Description

  • An excellent choice for general laboratory use
  • Effective for both liquid and solid mediums
  • Offer light weight, resilience, and compressibility
  • Chemically inert and moisture resistant
  • Sold in packs of 100
Specifications

Specifications

Cork Stopper
100 Pk.
Tapered
Non-sterile
1
13 mm
Cork
8 mm
Not Autoclavable
Tan
1
11 mm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit