Learn More
Chemglass Life Sciences Cork Boring Set, Plated Brass, Set of 12, 4-20mm
SDP
Supplier:  Chemglass Life Sciences CG3026C10
Encompass_Preferred
Cork Boring Set, Plated Brass, Set of 12, 4-20mm. For boring holes in rubber stoppers or corks. Plated brass tubing forms the borer and each borer has an individual handle. A punch is provided with each set. The 12-borer set covers range 4 to 20 mm.
Catalog No. 50-153-1487