Learn More
Cole-Parmer Raven Environmental B-10104-24 Coretaker Polycarbonate Sludge Sampler, 24-ft
Supplier:  Cole-Parmer 0546562
Encompass_Preferred
Two, rigid 8-ft lengths of clear, 1-1/4"dia polycarbonate tubing will not turn cloudy from sunlight or become brittle in cold weather like acrylic sludge samplers. The design offers a Slip-Loc quick-disconnect coupler and twin double o-ring seals to ensure a rugged, liquid-tight connection. Coretaker features a check valve for sample release; soft foam, floating lanyard handle; and pre-numbered yellow 1-ft markers.
Catalog No. 50-632-415
Explore available promotions