Learn More
Cole-Parmer Raven Environmental B-10104-16-REDESIGNED Coretaker Polycarbonate Sludge Sampler, 16-ft
Supplier: Cole-Parmer 0546560
This sampler is ideal for direct measurement of sludge blanket thickness by core sample extraction. It floats for easy retrieval, and there arent any threaded couplers, since it uses a Slip-Loc quick-disconnect design. It comes with a lifetime, no breakage guarantee.