Learn More
Perkin Elmer US LLC Copper Sample Pans and Covers - Qty. 200 ea.
Supplier: Perkin Elmer US LLC 03190026
PerkinElmer supplies specialty pans and covers for applications that require high temperatures or large sample volumes. With a maximum pressure threshold of ambient, most of these pans do not require a crimper. These pans and covers are compatible with the the DSC 4000/6000/8000/8500, DSC 6, DSC 7, Jade DSC, Diamond DSC, PYRIS 1 DSC, and PYRIS 6 DSC.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.