Learn More
Enzo Life Sciences COP9 signalosome (human), (purified) (20 µg)
SDP
Supplier:  Enzo Life Sciences BMLPW94250020
Encompass_Preferred
Alternative name: CSN. Formulation: Liquid. In 20mM TRIS, pH 7.2, containing 10% glycerol, 50mM KCl  and 1mM mercaptoethanol. Source: Isolated from human erythrocytes. Long Term Storage: -80°C.
Catalog No. 50-201-1687