Learn More
IMEB, Inc Pathology Bone Band Saw Optional Cooling System (Required for Diamond Blades)
Supplier: IMEB, Inc C82211
Optional Cooling System (Required for Diamond Blades). IMEB Inc. Bone Band Saw sold separately. Cooling system is for the IMEB, Inc. Bone Band Saw which is an efficient and economical way of cutting bone in laboratory. The bone band saw only 19" high and has a foot print of 17" x 13" of counter space. This saw has a separate water-cooled blade that eliminates dust and particles.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.