Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purus™ Controlled Environments Mat: PROMO

Removes particles from foot traffic and equipment wheels.

Supplier:  Purus™ PFM 3150 61 01PM

 View more versions of this product

Catalog No. 19-125-940PM

Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.

View alternate product


Description

Description

  • Each layer is coated with an adhesive to capture soil and contain it until the layer is peeled away and discarded
  • Numbered tabs ensure one layer at a time is removed and indicates the number of remaining layers
  • Non-skid backing is applied to the bottom of the frame to provide stability on a variety of floor surfaces
  • Frame mat can be easily relocated to high-traffic areas and removed during routine floor maintenanc
  • Protect a facility from dust, dirt and contamination
  • 30- or 60-layered mats applied to reusable polystyrene frames
  • Length 61cm (24 in.)
  • Width 76.2cm (30 in.)
Specifications

Specifications

61 x 76.2 cm (24 x 30 in.)
60 Sheets/Mat
Double White frame
White
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit