Learn More
Audit™ MicroControls™ Control DROP LQ Urine Dipstick
SDP
A bi-level human source material urine control fortified to target levels with compounds that produce the desired reaction when tested by the methods indicated.
Supplier:  Audit™ MicroControls™ K064M10
Catalog No. 22-157-143
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
2°C to 8°C
10 x 5 mL
Liquid